siRNA Details
Gene Symbol: MED8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001653.2 | NP_001001653.1 | 7 | 2001 | ||
NM_052877.4 | NP_443109.2 | 8 | 1465 | |||
NM_201542.4 | NP_963836.2 | 7 | 1958 | |||
GenBank mRNA | BC010543.1 | - | 7 | 1406 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
