siRNA Details
Gene Symbol: CMTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052999.3 | NP_443725.3 | 3 | 743 | ||
NM_181268.2 | NP_851785.2 | 3 | 743 | |||
NM_181269.2 | NP_851786.1 | 3 | 448 | |||
NM_181270.2 | NP_851787.1 | 3 | 448 | |||
NM_181271.2 | NP_851788.1 | 2 | 307 | |||
NM_181272.2 | NP_851789.1 | 2 | 289 | |||
NM_181283.2 | NP_851800.1 | 2 | 289 | |||
GenBank mRNA | AF278576.1 | - | 3 | 363 | ||
AF278577.1 | - | 3 | 363 | |||
AY174118.1 | - | 3 | 452 | |||
AY174119.1 | - | 3 | 452 | |||
AY174120.1 | - | 2 | 311 | |||
AY174121.1 | - | 2 | 293 | |||
AY174122.1 | - | 2 | 293 | |||
AY174123.1 | - | 2 | 448 | |||
AY174124.1 | - | 2 | 448 | |||
AY174125.1 | - | 4 | 586 | |||
AY174126.1 | - | 4 | 586 | |||
AY174128.1 | - | 3 | 416 | |||
AY174129.1 | - | 4 | 439 | |||
AY174130.1 | - | 4 | 611 | |||
AY174131.1 | - | 2 | 268 | |||
AY174132.1 | - | 2 | 268 | |||
BC057852.1 | - | 3 | 743 | 5273108 | ||
DQ309766.1 | - | 4 | 577 | |||
DQ355985.1 | - | 2 | 299 |
Gene Symbol: CKLF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA234074.1 | - | 2 | 240 |
Gene Symbol: CKLF-CMTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202509.1 | NP_001189438.1 | 2 | 316 | ||
NM_001204098.1 | NP_001191027.1 | 3 | 412 | |||
NM_001204099.1 | NP_001191028.1 | 3 | 475 | |||
GenBank mRNA | BM929895.1 | - | 2 | 264 | ||
FY211334.1 | - | 3 | 296 | |||
FY212695.1 | - | 3 | 359 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
