siRNA Details
Gene Symbol: GNRHR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002328.4 | 1 | 837 | |||
NR_104033.1 | 2 | 246 | ||||
NR_104034.1 | 2 | 400 | ||||
GenBank mRNA | AB239331.1 | - | 2 | 400 | ||
AK310736.1 | - | 2 | 727 | |||
AY077708.1 | - | 1 | 837 | |||
AY081843.1 | - | 2 | 246 | |||
AY233133.1 | - | 2 | 346 | |||
DQ266410.1 | - | 2 | 400 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
