siRNA Details
Gene Symbol: CMTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052999.3 | NP_443725.3 | 4 | 830 | ||
NM_181268.2 | NP_851785.2 | 4 | 814 | |||
NM_181269.2 | NP_851786.1 | 4 | 535 | |||
NM_181270.2 | NP_851787.1 | 4 | 519 | |||
NM_181271.2 | NP_851788.1 | 3 | 394 | |||
NM_181272.2 | NP_851789.1 | 3 | 376 | |||
NM_181283.2 | NP_851800.1 | 3 | 360 | |||
NM_181296.2 | NP_851813.1 | 2 | 261 | |||
GenBank mRNA | AF278576.1 | - | 4 | 434 | ||
AF278577.1 | - | 4 | 450 | |||
AY174118.1 | - | 4 | 539 | |||
AY174119.1 | - | 4 | 523 | |||
AY174120.1 | - | 3 | 398 | |||
AY174121.1 | - | 3 | 380 | |||
AY174122.1 | - | 3 | 364 | |||
AY174123.1 | - | 3 | 535 | |||
AY174124.1 | - | 3 | 519 | |||
AY174125.1 | - | 5 | 673 | |||
AY174126.1 | - | 5 | 657 | |||
AY174128.1 | - | 4 | 503 | |||
AY174129.1 | - | 5 | 526 | |||
AY174130.1 | - | 5 | 698 | |||
AY174131.1 | - | 3 | 355 | |||
AY174132.1 | - | 3 | 339 | |||
AY174133.1 | - | 2 | 265 | |||
AY174134.1 | - | 4 | 585 | |||
AY174135.1 | - | 5 | 763 | |||
AY174136.1 | - | 5 | 747 | |||
AY174137.1 | - | 4 | 565 | |||
AY174138.1 | - | 4 | 549 | |||
BC057852.1 | - | 4 | 830 | 5273108 | ||
DQ309766.1 | - | 5 | 648 | |||
DQ355985.1 | - | 3 | 386 |
Gene Symbol: CKLF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA234074.1 | - | 3 | 327 |
Gene Symbol: CKLF-CMTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202509.1 | NP_001189438.1 | 3 | 403 | ||
NM_001204098.1 | NP_001191027.1 | 4 | 483 | |||
NM_001204099.1 | NP_001191028.1 | 4 | 562 | |||
GenBank mRNA | FY211334.1 | - | 4 | 367 | ||
FY212695.1 | - | 4 | 446 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
