siRNA Details
Gene Symbol: STAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003150.3 | NP_003141.2 | 24 | 4060 | ||
NM_139276.2 | NP_644805.1 | 24 | 4085 | |||
NM_213662.1 | NP_998827.1 | 24 | 3927 | |||
XM_011525145.2 | XP_011523447.1 | 24 | 4076 | |||
XM_011525146.2 | XP_011523448.1 | 24 | 4026 | |||
XM_017024973.1 | XP_016880462.1 | 24 | 4023 | |||
GenBank mRNA | BC008044.2 | - | 1 | 557 | ||
BC067119.1 | - | 1 | 205 | |||
BC107775.1 | - | 1 | 412 | |||
CF454565.1 | - | 1 | 203 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
