siRNA Details
Gene Symbol: GNRHR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002328.4 | 2 | 960 | |||
NR_104033.1 | 3 | 369 | ||||
GenBank mRNA | AY077708.1 | - | 2 | 960 | ||
AY081843.1 | - | 3 | 369 | |||
AY233133.1 | - | 3 | 469 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
