siRNA Details
Gene Symbol: IL22RA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052962.2 | NP_443194.1 | 6, 7 | 951 | ||
NM_181309.1 | NP_851826.1 | 5, 6 | 855 | |||
GenBank mRNA | AJ297262.1 | - | 4, 5 | 575 | ||
AJ313161.1 | - | 5, 6 | 857 | |||
AJ313162.1 | - | 6, 7 | 953 | |||
AY040566.1 | - | 5, 6 | 828 | |||
AY040567.1 | - | 6, 7 | 924 | |||
AY044429.1 | - | 5, 6 | 790 | |||
AY358111.1 | - | 5, 6 | 853 | |||
AY358737.1 | - | 6, 7 | 887 | |||
BC125167.1 | - | 4, 5 | 617 | 40121609 | ||
BC125168.1 | - | 4, 5 | 617 | 40121612 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
