siRNA Details
Gene Symbol: LRG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052972.2 | NP_443204.1 | 2 | 1325 | ||
GenBank mRNA | AB073607.1 | - | 2 | 1337 | ||
AF403428.1 | - | 2 | 1325 | |||
AK027382.1 | - | 2 | 1337 | |||
BC034389.1 | - | 2 | 1326 | 5187278 | ||
BC070198.1 | - | 2 | 1382 | 4692618 |
Gene Symbol: GSDMB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011525004.1 | XP_011523306.1 | 1 | 515 | ||
XM_011525006.1 | XP_011523308.1 | 1 | 515 | |||
XM_011525007.2 | XP_011523309.1 | 1 | 515 | |||
XM_011525009.1 | XP_011523311.1 | 1 | 515 | |||
XM_011525011.1 | XP_011523313.1 | 1 | 515 | |||
XM_011525012.1 | XP_011523314.1 | 1 | 515 | |||
XM_011525013.1 | XP_011523315.1 | 1 | 515 | |||
XM_011525015.1 | XP_011523317.1 | 1 | 515 | |||
XM_017024848.1 | XP_016880337.1 | 1 | 515 | |||
XM_017024849.1 | XP_016880338.1 | 1 | 515 | |||
XM_017024850.1 | XP_016880339.1 | 1 | 515 | |||
XM_017024851.1 | XP_016880340.1 | 1 | 515 | |||
XM_017024852.1 | XP_016880341.1 | 1 | 515 | |||
XM_017024853.1 | XP_016880342.1 | 1 | 515 | |||
XM_017024854.1 | XP_016880343.1 | 1 | 515 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
