siRNA Details
Gene Symbol: FAM3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_058186.3 | NP_478066.3 | 2 | 184 | ||
XM_011529648.2 | XP_011527950.2 | 2 | 527 | |||
XM_011529649.2 | XP_011527951.1 | 2 | 101 | |||
GenBank mRNA | AF375989.1 | - | 2 | 104 | ||
AF494379.1 | - | 2 | 38 | |||
AJ409094.1 | - | 3 | 221 | |||
AK125528.1 | - | 2 | 186 | |||
AK302095.1 | - | 2 | 80 | |||
AY358459.1 | - | 2 | 52 | |||
BC057829.1 | - | 2 | 106 | 30338490 | ||
BI712971.1 | - | 2 | 146 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
