siRNA Details
Gene Symbol: MYO18A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_078471.3 | NP_510880.2 | 42 | 6982 | ||
NM_203318.1 | NP_976063.1 | 41 | 6937 | |||
GenBank mRNA | AB177858.1 | - | 41 | 6801 | ||
AB177860.1 | - | 41 | 5889 | |||
AF252258.1 | - | 4 | 1235 | |||
AK122870.1 | - | 8 | 2073 | |||
AY703984.1 | - | 41 | 5889 | |||
BC039612.1 | - | 40 | 6930 | 6045482 | ||
CR933614.1 | - | 41 | 6911 | |||
D86970.2 | - | 42 | 7005 |
Gene Symbol: TIAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004740.3 | NP_004731.2 | 1 | 1499 | ||
GenBank mRNA | AF105277.1 | - | 1 | 208 | ||
AK054958.1 | - | 1 | 1499 | |||
AK126861.1 | - | 1 | 3196 | |||
BC047669.1 | - | 1 | 796 | 6056136 | ||
BC058075.1 | - | 1 | 482 | |||
BC131561.1 | - | 1 | 696 | 40109188 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
