siRNA Details
Gene Symbol: ZNF197 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005264783.4 | XP_005264840.1 | 6 | 2785 | ||
GenBank mRNA | AF011573.1 | - | 5 | 2572 | ||
AK314196.1 | - | 6 | 2753 | |||
L32163.1 | - | 1 | 1386 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
