siRNA Details
Gene Symbol: B3GALT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080605.3 | NP_542172.2 | 1 | 1530 | ||
GenBank mRNA | BG290330.1 | - | 175 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
