siRNA Details
Gene Symbol: FDX1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031734.3 | NP_001026904.2 | 4, 5 | 422 | ||
GenBank mRNA | AK300568.1 | - | 4, 5 | 415 | ||
BC063460.1 | - | 4, 5 | 406 | 5179971 | ||
DA243025.1 | - | 4, 5 | 422 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
