siRNA Details
Gene Symbol: MACROD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080676.5 | NP_542407.2 | 2 | 404 | ||
XM_017027677.1 | XP_016883166.1 | 2 | 428 | |||
GenBank mRNA | AK131348.1 | - | 2 | 535 | ||
BC101216.1 | - | 2 | 185 | |||
BC101217.1 | - | 2 | 185 | |||
BC101218.1 | - | 2 | 185 | |||
BC101219.1 | - | 2 | 185 | |||
BC126936.1 | - | 2 | 185 | |||
BC128036.1 | - | 2 | 185 | |||
BX281637.1 | - | 2 | 281 | |||
DB125529.1 | - | 2 | 535 | |||
DB495787.2 | - | 404 | ||||
EF159161.1 | - | 140 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
