siRNA Details
Gene Symbol: TECTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_058222.2 | NP_478129.1 | 2 | 286 | ||
XM_017016604.1 | XP_016872093.1 | 2 | 280 | |||
GenBank mRNA | AF312827.1 | - | 2 | 207 | ||
BC113497.1 | - | 2 | 286 | 8322549 | ||
BC113499.1 | - | 2 | 286 | 8322551 |
