siRNA Details
Gene Symbol: SP140 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278451.1 | NP_001265380.1 | 7 | 820 | ||
NM_001278453.1 | NP_001265382.1 | 7 | 811 | |||
NM_007237.4 | NP_009168.4 | 7 | 820 | |||
XM_005246252.2 | XP_005246309.1 | 7 | 775 | |||
XM_005246254.2 | XP_005246311.1 | 7 | 784 | |||
XM_005246255.2 | XP_005246312.1 | 7 | 784 | |||
XM_005246256.2 | XP_005246313.1 | 7 | 784 | |||
XM_006712223.3 | XP_006712286.1 | 7 | 784 | |||
XM_011510515.1 | XP_011508817.1 | 7 | 784 | |||
XM_011510516.1 | XP_011508818.1 | 7 | 784 | |||
XM_011510517.2 | XP_011508819.1 | 9 | 1648 | |||
XM_011510518.2 | XP_011508820.1 | 8 | 920 | |||
XM_011510519.1 | XP_011508821.1 | 7 | 784 | |||
XM_017003239.1 | XP_016858728.1 | 7 | 775 | |||
XM_017003241.1 | XP_016858730.1 | 7 | 784 | |||
XM_017003243.1 | XP_016858732.1 | 7 | 775 | |||
XM_017003245.1 | XP_016858734.1 | 7 | 775 | |||
XM_017003248.1 | XP_016858737.1 | 7 | 775 | |||
XM_017003252.1 | XP_016858741.1 | 7 | 784 | |||
GenBank mRNA | BC038091.1 | - | 7 | 791 | ||
BC105960.1 | - | 7 | 805 | 40035752 | ||
U36499.1 | - | 7 | 821 | |||
U36500.1 | - | 7 | 821 | |||
U63420.1 | - | 7 | 802 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
