siRNA Details
Gene Symbol: C21orf58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006724018.3 | XP_006724081.1 | 7, 8 | 1969 | ||
GenBank mRNA | AY039244.1 | - | 6, 7 | 707 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
