siRNA Details
Gene Symbol: MYO18A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_078471.3 | NP_510880.2 | 42 | 7250 | ||
NM_203318.1 | NP_976063.1 | 41 | 7205 | |||
GenBank mRNA | AB177858.1 | - | 41 | 7069 | ||
AB177860.1 | - | 41 | 6157 | |||
AF252258.1 | - | 4 | 1503 | |||
AK122870.1 | - | 8 | 2341 | |||
AK303522.1 | - | 29 | 3627 | |||
AY703984.1 | - | 41 | 6157 | |||
BC039612.1 | - | 40 | 7198 | 6045482 | ||
CR933614.1 | - | 41 | 7179 | |||
D86970.2 | - | 42 | 7273 |
Gene Symbol: TIAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004740.3 | NP_004731.2 | 1 | 1767 | ||
GenBank mRNA | AF105277.1 | - | 1 | 476 | ||
AK054958.1 | - | 1 | 1767 | |||
AK126861.1 | - | 1 | 3464 | |||
BC047669.1 | - | 1 | 1064 | 6056136 | ||
BC058075.1 | - | 1 | 750 | |||
BC131561.1 | - | 1 | 964 | 40109188 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
