siRNA Details
Gene Symbol: TMEM41A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080652.3 | NP_542383.1 | 5 | 1062 | ||
XM_017007437.1 | XP_016862926.1 | 4 | 900 | |||
GenBank mRNA | AK290878.1 | - | 5 | 1040 | ||
AL832782.1 | - | 2 | 521 | |||
AY358547.1 | - | 5 | 1046 | |||
BC008043.2 | - | 5 | 1020 | 3838280 | ||
BC019884.1 | - | 5 | 1009 | 4991416 | ||
BX647128.1 | - | 3 | 712 |
