siRNA Details
Gene Symbol: ASB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201583.1 | NP_001188512.1 | 1 | 154 | ||
NM_080873.2 | NP_543149.1 | 1 | 154 | |||
GenBank mRNA | AF425642.1 | - | 1 | 135 | ||
BC069340.1 | - | 1 | 135 | 7262279 | ||
BC103873.1 | - | 1 | 135 | |||
BC103874.1 | - | 1 | 135 | 40004166 | ||
BC103875.1 | - | 1 | 135 | 40004167 | ||
BX537857.1 | - | 1 | 154 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
