siRNA Details
Gene Symbol: UNG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003362.3 | NP_003353.1 | 3 | 631 | ||
NM_080911.2 | NP_550433.1 | 4 | 535 | |||
GenBank mRNA | AK291341.1 | - | 3 | 555 | ||
AK299416.1 | - | 4 | 475 | |||
AK313552.1 | - | 4 | 532 | |||
BC015205.1 | - | 3 | 514 | 3913667 | ||
BC050634.2 | - | 3 | 525 | 6500377 | ||
BM928006.1 | - | 586 | ||||
BQ950839.1 | - | 4 | 546 | |||
CR541673.1 | - | 3 | 422 | |||
X15653.1 | - | 3 | 528 | |||
Y09008.1 | - | 4 | 519 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
