siRNA Details
Gene Symbol: PAGE2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015038.2 | NP_001015038.1 | 2 | 118 | ||
XM_011530785.2 | XP_011529087.1 | 3 | 534 | |||
XM_011530786.2 | XP_011529088.1 | 2 | 201 | |||
XM_011530787.2 | XP_011529089.1 | 2 | 181 | |||
XM_017029513.1 | XP_016885002.1 | 2 | 138 | |||
GenBank mRNA | BC130359.1 | - | 2 | 77 | 40146390 | |
BM782405.1 | - | 2 | 103 |
Gene Symbol: PAGE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207339.3 | NP_997222.1 | 2 | 111 | ||
XM_017029353.1 | XP_016884842.1 | 2 | 140 | |||
GenBank mRNA | BC054022.1 | - | 2 | 105 | 6603202 | |
BX113994.1 | - | 2 | 92 |
Gene Symbol: PAGE5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013435.2 | NP_001013453.1 | 2 | 162 | ||
NM_130467.4 | NP_569734.2 | 2 | 361 | |||
GenBank mRNA | AJ344352.1 | - | 2 | 331 | ||
BC009230.2 | - | 2 | 350 | 3955765 | ||
BC110803.1 | - | 2 | 96 | 3922538 | ||
BP429046.1 | - | 2 | 166 | |||
BQ437151.1 | - | 2 | 150 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
