siRNA Details
Gene Symbol: TPTE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130785.3 | NP_570141.3 | 5, 6 | 499 | ||
NR_073487.1 | 5, 6 | 499 | ||||
GenBank mRNA | AJ421032.1 | - | 5, 6 | 499 | ||
BC128147.1 | - | 5, 6 | 481 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
