siRNA Details
Gene Symbol: PAGE5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013435.2 | NP_001013453.1 | 3 | 294 | ||
NM_130467.4 | NP_569734.2 | 3 | 493 | |||
GenBank mRNA | AJ344352.1 | - | 3 | 463 | ||
BC009230.2 | - | 3 | 482 | 3955765 | ||
BC110803.1 | - | 3 | 228 | 3922538 | ||
BP429046.1 | - | 3 | 298 | |||
BQ437151.1 | - | 3 | 282 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
