siRNA Details
Gene Symbol: CHST14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130468.3 | NP_569735.1 | 1 | 1308 | ||
GenBank mRNA | AB066595.1 | - | 1 | 1271 | ||
AF282905.1 | - | 1 | 1107 | |||
AF401222.1 | - | 1 | 1055 | |||
AK074739.1 | - | 1 | 1063 | |||
AY358446.1 | - | 1 | 952 | |||
BC023653.2 | - | 1 | 942 | |||
BC049214.1 | - | 1 | 1255 | 5518321 | ||
BC053633.1 | - | 1 | 1214 | 6501263 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
