siRNA Details
Gene Symbol: DTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318043.1 | NP_001304972.1 | 5 | 1233 | ||
GenBank mRNA | AK074304.1 | - | 5 | 1163 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
