siRNA Details
Gene Symbol: SLC22A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080866.2 | NP_543142.2 | 5 | 1159 | ||
XM_017017159.1 | XP_016872648.1 | 5 | 2077 | |||
XM_017017160.1 | XP_016872649.1 | 4 | 736 | |||
GenBank mRNA | AB062418.1 | - | 5 | 992 | ||
AB074812.1 | - | 5 | 1140 | |||
AF440402.1 | - | 5 | 1127 | |||
AJ295270.1 | - | 5 | 951 | |||
AK074246.1 | - | 5 | 1159 | |||
BC022379.1 | - | 4 | 1004 | |||
BC126288.1 | - | 5 | 1041 | 8992004 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
