siRNA Details
Gene Symbol: DUSP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142314.1 | NP_001135786.1 | 3 | 1013 | ||
NM_001321519.1 | NP_001308448.1 | 4 | 1048 | |||
NM_080876.3 | NP_543152.1 | 4 | 1166 | |||
NR_135688.1 | 3 | 1119 | ||||
GenBank mRNA | AB063186.1 | - | 4 | 1171 | ||
AB063187.1 | - | 3 | 1018 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
