siRNA Details
Gene Symbol: RAB40A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080879.2 | NP_543155.2 | 3 | 1185 | ||
XM_005262083.4 | XP_005262140.1 | 2 | 1662 | |||
XM_011530872.2 | XP_011529174.1 | 2 | 1659 | |||
GenBank mRNA | AF132748.1 | - | 1 | 908 | ||
AF422143.1 | - | 3 | 1188 | |||
AK056859.1 | - | 1 | 2962 | |||
BC074854.2 | - | 1 | 874 | 30915461 | ||
BC074855.2 | - | 1 | 874 | 30915224 | ||
BC113501.1 | - | 1 | 926 | 8322553 | ||
BC117232.1 | - | 1 | 926 | 40125783 |
Gene Symbol: RAB40AL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031834.1 | NP_001027004.1 | 1 | 893 | ||
GenBank mRNA | BC101169.1 | - | 1 | 893 | 40016064 | |
BC101170.2 | - | 1 | 893 | 40016065 | ||
BC101171.1 | - | 1 | 893 | 40016067 | ||
BC101172.1 | - | 1 | 893 | 40016069 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
