siRNA Details
Gene Symbol: RAB24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109789.1 | 6 | 1006 | |||
NR_109790.1 | 5 | 1092 | ||||
GenBank mRNA | AF258583.1 | - | 5 | 836 | ||
AK021761.1 | - | 4 | 1123 | |||
AK308155.1 | - | 6 | 961 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
