siRNA Details
Gene Symbol: FAM181A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207071.1 | NP_001194000.1 | 2 | 484 | ||
NM_001207072.1 | NP_001194001.1 | 2 | 625 | |||
NM_001207073.1 | NP_001194002.1 | 2 | 495 | |||
NM_001207074.1 | NP_001194003.1 | 2 | 441 | |||
NM_138344.4 | NP_612353.3 | 3 | 807 | |||
XM_011537297.1 | XP_011535599.1 | 2 | 458 | |||
GenBank mRNA | AK315393.1 | - | 2 | 495 | ||
AY255795.1 | - | 2 | 461 | |||
BC009073.1 | - | 2 | 625 | 3856004 | ||
CX870424.1 | - | 2 | 441 | |||
DB058808.1 | - | 2 | 484 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
