siRNA Details
Gene Symbol: SAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320845.1 | NP_001307774.1 | 4 | 521 | ||
NM_133491.4 | NP_597998.1 | 4 | 504 | |||
XM_017024073.1 | XP_016879562.1 | 4 | 521 | |||
GenBank mRNA | AF348524.1 | - | 4 | 263 | ||
BC011751.2 | - | 4 | 474 | 3534187 | ||
BC065922.1 | - | 1 | 813 | |||
BC095466.1 | - | 4 | 459 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
