siRNA Details
Gene Symbol: TMEM129 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127266.1 | NP_001120738.1 | 4 | 1704 | ||
NM_138385.3 | NP_612394.1 | 3 | 1544 | |||
XM_005248038.4 | XP_005248095.1 | 4 | 1576 | |||
XM_005248039.4 | XP_005248096.1 | 4 | 1145 | |||
XM_011513594.2 | XP_011511896.1 | 4 | 1154 | |||
XM_017008834.1 | XP_016864323.1 | 4 | 1441 | |||
GenBank mRNA | AK098466.1 | - | 3 | 559 | ||
AK303880.1 | - | 4 | 1017 | |||
AK310320.1 | - | 3 | 616 | |||
BC009331.2 | - | 3 | 1544 | |||
BC018921.2 | - | 3 | 1544 | |||
BC126370.1 | - | 5 | 1594 | 8992086 |
