siRNA Details
Gene Symbol: SRPK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003137.4 | NP_003128.3 | 3, 4 | 325 | ||
NR_034069.1 | 3, 4 | 353 | ||||
GenBank mRNA | AJ318054.1 | - | 2, 3 | 723 | ||
AK292277.1 | - | 3, 4 | 270 | |||
AK299591.1 | - | 3, 4 | 313 | |||
AK312712.1 | - | 3, 4 | 217 | |||
BC038292.1 | - | 3, 4 | 290 | 4824261 | ||
BG772464.1 | - | 3, 4 | 333 | |||
BX648663.1 | - | 3, 4 | 242 | |||
BX648697.1 | - | 3, 4 | 243 | |||
DB233331.1 | - | 3, 4 | 325 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_016795, AK017187, AJ224115, AK161346, AB012290, and others.
Gene Ontology Categories:
