siRNA Details
Gene Symbol: GPT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142466.2 | NP_001135938.1 | 12 | 3337 | ||
NM_133443.3 | NP_597700.1 | 12 | 3361 | |||
XM_017023790.1 | XP_016879279.1 | 9 | 2834 | |||
GenBank mRNA | AK090957.1 | - | 3 | 2044 | ||
AK094971.1 | - | 12 | 3260 | |||
AL833351.1 | - | 3 | 2599 | |||
AY029173.2 | - | 12 | 3344 | |||
BC051364.1 | - | 3 | 1992 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
