siRNA Details
Gene Symbol: FAM122A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138333.4 | NP_612206.4 | 1 | 1118 | ||
GenBank mRNA | AA418448.1 | - | 1 | 230 | ||
AK126379.1 | - | 1 | 1059 | |||
BC013062.1 | - | 1 | 1069 | 2901027 | ||
BE778779.1 | - | 111 |
