siRNA Details
Gene Symbol: MCU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270679.1 | NP_001257608.1 | 6, 7 | 827 | ||
NM_001270680.1 | NP_001257609.1 | 6, 7 | 1167 | |||
NM_138357.2 | NP_612366.1 | 6, 7 | 890 | |||
NR_073062.1 | 8, 9 | 1055 | ||||
XM_017016882.1 | XP_016872371.1 | 6, 7 | 1090 | |||
GenBank mRNA | AK025699.1 | - | 2, 3 | 232 | ||
AK075439.1 | - | 6, 7 | 877 | |||
AK128016.1 | - | 6, 7 | 1167 | |||
AK301475.1 | - | 8, 9 | 1036 | |||
AK315519.1 | - | 6, 7 | 873 | |||
BC010682.1 | - | 6, 7 | 784 | |||
BC034235.1 | - | 6, 7 | 890 | 5296557 |

More Information
Other Species Targeted

Gene Ontology Categories:
