siRNA Details
Gene Symbol: ADPRHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138430.4 | NP_612439.2 | 1 | 167 | ||
GenBank mRNA | AJ313429.1 | - | 1 | 167 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
