siRNA Details
Gene Symbol: ZADH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306093.1 | NP_001293022.1 | 2 | 2525 | ||
NM_175907.5 | NP_787103.1 | 2 | 2703 | |||
GenBank mRNA | BC010734.1 | - | 1 | 1190 | ||
BC078661.1 | - | 2 | 2514 | 5575156 | ||
BX647730.1 | - | 2 | 2528 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
