siRNA Details
Gene Symbol: ADAMTS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_197941.3 | NP_922932.2 | 25 | 6644 | ||
NR_135689.1 | 26 | 6795 | ||||
XM_011543113.2 | XP_011541415.1 | 25 | 6659 | |||
XM_011543114.2 | XP_011541416.1 | 25 | 6238 | |||
XM_011543115.2 | XP_011541417.1 | 24 | 6547 | |||
XM_011543116.2 | XP_011541418.1 | 23 | 6406 | |||
XM_011543117.1 | XP_011541419.1 | 23 | 5997 | |||
XM_011543121.2 | XP_011541423.2 | 21 | 5025 | |||
XM_011543123.2 | XP_011541425.1 | 19 | 4763 | |||
XM_011543124.2 | XP_011541426.1 | 16 | 4547 | |||
XM_011543125.1 | XP_011541427.1 | 15 | 4402 | |||
GenBank mRNA | AB209685.1 | - | 26 | 6292 | ||
AY692423.1 | - | 6 | 3247 | |||
AY692424.1 | - | 25 | 6602 | |||
BC010563.1 | - | 1 | 795 | |||
BC071654.1 | - | 1 | 1048 | |||
CR749356.1 | - | 7 | 3379 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
