siRNA Details
Gene Symbol: PRELID2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138492.5 | NP_612501.3 | 4, 5 | 532 | ||
NM_205846.2 | NP_995318.1 | 3, 4 | 329 | |||
XM_017009135.1 | XP_016864624.1 | 3, 4 | 329 | |||
GenBank mRNA | AK125215.1 | - | 3, 4 | 264 | ||
BC012033.1 | - | 4, 5 | 499 | 4476359 | ||
BC028175.1 | - | 246 | ||||
BQ645893.1 | - | 3, 4 | 299 | |||
BU167299.1 | - | 3, 4 | 191 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
