siRNA Details
Gene Symbol: NOXRED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113475.2 | NP_001106946.1 | 2 | 409 | ||
XM_005267330.3 | XP_005267387.1 | 3 | 1034 | |||
XM_011536427.2 | XP_011534729.1 | 3 | 1047 | |||
XM_011536428.2 | XP_011534730.1 | 3 | 294 | |||
XM_017020969.1 | XP_016876458.1 | 2 | 409 | |||
XM_017020970.1 | XP_016876459.1 | 2 | 409 | |||
GenBank mRNA | AK057371.1 | - | 2 | 410 | ||
BC066965.1 | - | 2 | 389 | 4373886 | ||
BI829015.1 | - | 3 | 301 | |||
DB021944.1 | - | 2 | 410 |
