siRNA Details
Gene Symbol: CCDC74A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258305.1 | NP_001245234.1 | 1, 2 | 394 | ||
NM_001258306.1 | NP_001245235.1 | 1, 2 | 394 | |||
NM_138770.2 | NP_620125.1 | 1, 2 | 394 | |||
XM_005263837.1 | XP_005263894.1 | 1, 2 | 340 | |||
XM_017005235.1 | XP_016860724.1 | 1, 2 | 553 | |||
XM_017005239.1 | XP_016860728.1 | 1, 2 | 553 | |||
XM_017005240.1 | XP_016860729.1 | 1, 2 | 553 | |||
XM_017005245.1 | XP_016860734.1 | 1, 2 | 340 | |||
XM_017005246.1 | XP_016860735.1 | 1, 2 | 394 | |||
GenBank mRNA | AB299087.1 | - | 1, 2 | 309 | ||
AK309113.1 | - | 1, 2 | 553 | |||
BC016861.1 | - | 1, 2 | 309 | 3914851 | ||
BC063387.1 | - | 1, 2 | 331 | 5494806 | ||
CV023784.1 | - | 1, 2 | 257 | |||
DA375407.1 | - | 1, 2 | 394 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
