siRNA Details
Gene Symbol: CCDC148 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301685.1 | NP_001288614.1 | 3 | 547 | ||
NM_138803.3 | NP_620158.3 | 3 | 547 | |||
XM_005246319.3 | XP_005246376.1 | 4 | 434 | |||
XM_011510638.2 | XP_011508940.1 | 4 | 435 | |||
XM_017003392.1 | XP_016858881.1 | 3 | 547 | |||
XM_017003393.1 | XP_016858882.1 | 3 | 230 | |||
XM_017003394.1 | XP_016858883.1 | 2 | 380 | |||
XM_017003397.1 | XP_016858886.1 | 1 | 72 | |||
GenBank mRNA | AF458588.1 | - | 3 | 496 | ||
AK058105.1 | - | 4 | 409 | |||
AK309246.1 | - | 2 | 380 | |||
BC015395.1 | - | 3 | 496 | |||
BC107597.1 | - | 3 | 360 | 40025920 |
