siRNA Details
Gene Symbol: TGIF2LX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF497480.1 | - | 1 | 613 | ||
BC101144.2 | - | 1 | 618 | |||
BC101145.1 | - | 1 | 618 | |||
BC117398.1 | - | 2 | 633 | 40125949 | ||
BC117400.1 | - | 2 | 633 | 40125951 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
