siRNA Details
Gene Symbol: RPS4Y2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039567.2 | NP_001034656.1 | 3 | 329 | ||
GenBank mRNA | AF497481.2 | - | 3 | 329 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
