siRNA Details
Gene Symbol: MAPKAPK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003668.3 | NP_003659.2 | 3, 4 | 700 | ||
NM_139078.2 | NP_620777.1 | 3, 4 | 700 | |||
XM_017020125.1 | XP_016875614.1 | 3, 4 | 192 | |||
XM_017020132.1 | XP_016875621.1 | 3, 4 | 558 | |||
XM_017020133.1 | XP_016875622.1 | 3, 4 | 454 | |||
XM_017020134.1 | XP_016875623.1 | 3, 4 | 280 | |||
XM_017020135.1 | XP_016875624.1 | 3, 4 | 453 | |||
XM_017020136.1 | XP_016875625.1 | 3, 4 | 454 | |||
XM_017020137.1 | XP_016875626.1 | 3, 4 | 453 | |||
XM_017020139.1 | XP_016875628.1 | 3, 4 | 453 | |||
GenBank mRNA | AK122767.1 | - | 3, 4 | 950 | ||
AW780199.1 | - | 3, 4 | 454 | |||
BC000833.1 | - | 2, 3 | 143 | |||
BC041049.1 | - | 3, 4 | 687 | 4909021 | ||
BC047284.1 | - | 3, 4 | 368 | 4997201 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
