siRNA Details
Gene Symbol: MTFMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139242.3 | NP_640335.2 | 3, 4 | 576 | ||
XM_005254158.4 | XP_005254215.2 | 3, 4 | 718 | |||
GenBank mRNA | AK055688.1 | - | 3, 4 | 522 | ||
BC016630.1 | - | 3, 4 | 554 | 4707448 | ||
BC033687.1 | - | 3, 4 | 564 | 5196677 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
