siRNA Details
Gene Symbol: SPATA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139073.3 | NP_620712.2 | 5 | 833 | ||
XM_005246303.2 | XP_005246360.1 | 4 | 751 | |||
GenBank mRNA | AY032925.1 | - | 6 | 931 | ||
BC047704.1 | - | 6 | 776 | 5744971 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
