siRNA Details
Gene Symbol: CCDC148 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301685.1 | NP_001288614.1 | 7 | 1086 | ||
GenBank mRNA | AK058105.1 | - | 8 | 948 | ||
BC015395.1 | - | 7 | 1035 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
